Product Details

SNP ID
rs144434621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65573521 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCTACCCACCGGATGCGGGCCT[A/G]TCTGACGACGAGGAGCCTCCCGATG
Phenotype
MIM: 616128 MIM: 606044
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EHBP1L1 PubMed Links

Gene Details

Gene
EHBP1L1
Gene Name
EH domain binding protein 1 like 1
There are no transcripts associated with this gene.

Gene
FAM89B
Gene Name
family with sequence similarity 89 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098784.1 771 Missense Mutation ATC,GTC I154V NP_001092254.1
NM_001098785.1 771 Silent Mutation CTA,CTG L150L NP_001092255.1
NM_152832.2 771 Silent Mutation CTA,CTG L137L NP_690045.1
Gene
SSSCA1
Gene Name
Sjogren syndrome/scleroderma autoantigen 1
There are no transcripts associated with this gene.

Gene
SSSCA1-AS1
Gene Name
SSSCA1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details