Product Details

SNP ID
rs144687659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65598564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGATGAGGCCCAGGGGTGGTGA[A/G]CGTGGGGTGCCTGGGGTGCCAGGAG
Phenotype
MIM: 603940 MIM: 600050
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EHBP1L1 PubMed Links

Gene Details

Gene
EHBP1L1
Gene Name
EH domain binding protein 1 like 1
There are no transcripts associated with this gene.

Gene
KCNK7
Gene Name
potassium two pore domain channel subfamily K member 7
There are no transcripts associated with this gene.

Gene
MAP3K11
Gene Name
mitogen-activated protein kinase kinase kinase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002419.3 2764 Silent Mutation CGC,CGT R757R NP_002410.1

View Full Product Details