Product Details

SNP ID
rs144833208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:69054090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCTGTGGTCTTCATCGAAGATG[C/G]TATTCAGGTCGGTGGCACCTGCTCC
Phenotype
MIM: 612163
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TPCN2 PubMed Links

Gene Details

Gene
TPCN2
Gene Name
two pore segment channel 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139075.3 287 Missense Mutation GCT,GGT A56G NP_620714.2
XM_005273824.3 287 Missense Mutation GCT,GGT A56G XP_005273881.1
XM_005273826.3 287 Missense Mutation GCT,GGT A56G XP_005273883.1
XM_005273830.3 287 UTR 5 XP_005273887.1
XM_005273831.3 287 UTR 5 XP_005273888.1
XM_005273832.3 287 UTR 5 XP_005273889.1
XM_011544802.2 287 Missense Mutation CTA,GTA L2V XP_011543104.1
XM_011544805.1 287 Missense Mutation GCT,GGT A56G XP_011543107.1
XM_011544806.1 287 Missense Mutation GCT,GGT A56G XP_011543108.1
XM_011544807.2 287 UTR 5 XP_011543109.1
XM_011544808.2 287 UTR 5 XP_011543110.1
XM_017017328.1 287 Missense Mutation GCT,GGT A56G XP_016872817.1
XM_017017329.1 287 Missense Mutation GCT,GGT A56G XP_016872818.1
XM_017017330.1 287 UTR 5 XP_016872819.1
XM_017017331.1 287 Intron XP_016872820.1
XM_017017332.1 287 UTR 5 XP_016872821.1
XM_017017333.1 287 UTR 5 XP_016872822.1
XM_017017334.1 287 UTR 5 XP_016872823.1
XM_017017335.1 287 Intron XP_016872824.1
XM_017017336.1 287 Intron XP_016872825.1

View Full Product Details