Product Details

SNP ID
rs144833612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125893936 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGGGTGCTCAATTCGTCCCCTA[C/T]GTACAAAATGCTGGATCCGGGATTC
Phenotype
MIM: 607663 MIM: 610693 MIM: 616283
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX25 PubMed Links

Gene Details

Gene
DDX25
Gene Name
DEAD-box helicase 25
There are no transcripts associated with this gene.

Gene
HYLS1
Gene Name
HYLS1, centriolar and ciliogenesis associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134793.1 1393 Intron NP_001128265.1
NM_145014.2 1393 Intron NP_659451.1
XM_005271430.2 1393 Intron XP_005271487.1
XM_006718777.3 1393 Intron XP_006718840.1
XM_011542659.2 1393 Intron XP_011540961.1
XM_017017320.1 1393 Intron XP_016872809.1
XM_017017321.1 1393 Intron XP_016872810.1
Gene
PUS3
Gene Name
pseudouridylate synthase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271985.1 1393 Missense Mutation CAT,CGT H224R NP_001258914.1
NM_031307.3 1393 Missense Mutation CAT,CGT H432R NP_112597.3
XM_005271687.3 1393 Missense Mutation CAT,CGT H333R XP_005271744.1
XM_005271688.3 1393 UTR 3 XP_005271745.1

View Full Product Details