Product Details

SNP ID
rs144853634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:113316362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAATTTCTGCTTTAGAGAAGTGG[A/C]GTAGATAAATGATTGCTCTAAGTTC
Phenotype
MIM: 610732
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TTC12 PubMed Links

Gene Details

Gene
TTC12
Gene Name
tetratricopeptide repeat domain 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318533.1 Intron NP_001305462.1
NM_017868.3 Intron NP_060338.3
XM_005271604.3 Intron XP_005271661.1
XM_011542890.2 Intron XP_011541192.1
XM_017017943.1 Intron XP_016873432.1
XM_017017944.1 Intron XP_016873433.1
XM_017017945.1 Intron XP_016873434.1
XM_017017946.1 Intron XP_016873435.1
XM_017017947.1 Intron XP_016873436.1
XM_017017948.1 Intron XP_016873437.1
XM_017017949.1 Intron XP_016873438.1
XM_017017950.1 Intron XP_016873439.1

View Full Product Details