Product Details

SNP ID
rs144971465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:110579612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCTGAGTCCTGGAAGGGAGAAG[A/T]ACTACACCTTTGGGCTGACTGCACA
Phenotype
MIM: 609568
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP20 PubMed Links

Gene Details

Gene
ARHGAP20
Gene Name
Rho GTPase activating protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258415.1 3440 Missense Mutation ACT,TCT T1089S NP_001245344.1
NM_001258416.1 3440 Missense Mutation ACT,TCT T1086S NP_001245345.1
NM_001258417.1 3440 Missense Mutation ACT,TCT T1076S NP_001245346.1
NM_001258418.1 3440 Missense Mutation ACT,TCT T1076S NP_001245347.1
NM_020809.3 3440 Missense Mutation ACT,TCT T1112S NP_065860.2
XM_005271628.3 3440 Missense Mutation ACT,TCT T655S XP_005271685.1
XM_006718893.3 3440 Missense Mutation ACT,TCT T1112S XP_006718956.1

View Full Product Details