Product Details

SNP ID
rs145196976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67468062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCCCCATCCGGCTCCGGAGAGG[A/C]CTGGGCTCCATCCTCATCGTTCTCC
Phenotype
MIM: 608965
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CABP4 PubMed Links

Gene Details

Gene
CABP4
Gene Name
calcium binding protein 4
There are no transcripts associated with this gene.

Gene
TMEM134
Gene Name
transmembrane protein 134
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001078650.2 290 Missense Mutation GCC,TCC A69S NP_001072118.1
NM_001078651.2 290 Missense Mutation GCC,TCC A69S NP_001072119.1
NM_025124.3 290 Missense Mutation GCC,TCC A69S NP_079400.1
XM_006718693.2 290 Missense Mutation GCC,TCC A69S XP_006718756.1
XM_011545268.2 290 Missense Mutation GCC,TCC A69S XP_011543570.1
XM_017018356.1 290 Missense Mutation GCC,TCC A69S XP_016873845.1

View Full Product Details