Product Details

SNP ID
rs145255279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59422135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGGCATAGAATATGGACACCAC[A/C]TTGTCCCTGTTTAGGGAGTAGCTGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR5A2 PubMed Links

Gene Details

Gene
OR5A2
Gene Name
olfactory receptor family 5 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001954.1 819 Missense Mutation AAG,AAT K273N NP_001001954.1

View Full Product Details