Product Details

SNP ID
rs145373644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:800187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCGTCTGCGCCCTTCCTCTGCC[A/G]GGCAGCCTCAGCCTCCTCGGGCACC
Phenotype
MIM: 605247 MIM: 180530 MIM: 609302
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PANO1 PubMed Links

Gene Details

Gene
PANO1
Gene Name
proapoptotic nucleolar protein 1
There are no transcripts associated with this gene.

Gene
PIDD1
Gene Name
p53-induced death domain protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145886.3 3375 Missense Mutation CGG,TGG R740W NP_665893.2
NM_145887.3 3375 Missense Mutation CGG,TGG R723W NP_665894.2
XM_005253005.4 3375 Missense Mutation CGG,TGG R764W XP_005253062.1
XM_005253006.4 3375 Missense Mutation CGG,TGG R764W XP_005253063.1
XM_005253007.3 3375 Missense Mutation CGG,TGG R764W XP_005253064.1
XM_005253008.4 3375 Missense Mutation CGG,TGG R618W XP_005253065.1
XM_011520209.2 3375 Missense Mutation CGG,TGG R750W XP_011518511.1
XM_011520210.2 3375 Missense Mutation CGG,TGG R740W XP_011518512.1
XM_011520211.2 3375 Missense Mutation CGG,TGG R723W XP_011518513.1
XM_011520212.1 3375 Missense Mutation CGG,TGG R618W XP_011518514.1
XM_011520213.2 3375 Missense Mutation CGG,TGG R451W XP_011518515.1
XM_017017993.1 3375 Missense Mutation CGG,TGG R427W XP_016873482.1
Gene
RPLP2
Gene Name
ribosomal protein lateral stalk subunit P2
There are no transcripts associated with this gene.

Gene
SLC25A22
Gene Name
solute carrier family 25 member 22
There are no transcripts associated with this gene.

View Full Product Details