Product Details

SNP ID
rs145383958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:26994871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGACCTAGACGGGGAAGAGAGCTA[C/T]GGCAAGTACCTGCGGCGGGAGTCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FIBIN PubMed Links

Gene Details

Gene
FIBIN
Gene Name
fin bud initiation factor homolog (zebrafish)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203371.1 791 Silent Mutation TAC,TAT Y115Y NP_976249.1

View Full Product Details