Product Details

SNP ID
rs145432284
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102944347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGTATTCAAACTGTATGGGTCC[A/G]TTGAAAAAATAGATATAACCTATAA
Phenotype
MIM: 600108
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MMP13 PubMed Links

Gene Details

Gene
MMP13
Gene Name
matrix metallopeptidase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002427.3 1364 Silent Mutation AAC,AAT N445N NP_002418.1

View Full Product Details