Product Details

SNP ID
rs145457736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67507472 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTCCGTGTAGGTGCTCTGGGAG[C/G]CCTGGGCGCCGGGTGGCTTCATCGC
Phenotype
MIM: 608794
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDK2AP2 PubMed Links

Gene Details

Gene
CDK2AP2
Gene Name
cyclin dependent kinase 2 associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271849.1 852 Intron NP_001258778.1
NM_005851.4 852 Missense Mutation GCC,GGC A69G NP_005842.1
Gene
PITPNM1
Gene Name
phosphatidylinositol transfer protein membrane associated 1
There are no transcripts associated with this gene.

View Full Product Details