Product Details

SNP ID
rs145512424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3659258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTCTGCTTCTTGTGTCTGTGGG[C/T]CTCATGGAAGCACTTCAGGTATGGG
Phenotype
MIM: 601625 MIM: 606372
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ART1 PubMed Links

Gene Details

Gene
ART1
Gene Name
ADP-ribosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004314.2 146 Silent Mutation GGC,GGT G15G NP_004305.2
XM_011520114.2 146 Silent Mutation GGC,GGT G15G XP_011518416.1
XM_017017763.1 146 Silent Mutation GGC,GGT G15G XP_016873252.1
Gene
CHRNA10
Gene Name
cholinergic receptor nicotinic alpha 10 subunit
There are no transcripts associated with this gene.

View Full Product Details