Product Details

SNP ID
rs145623767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:70206397 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTACAAGAGGTTCAGCAGGCCC[A/G]TGACCTCCAGAACAGGAGTGGGGCC
Phenotype
MIM: 602457
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FADD PubMed Links

Gene Details

Gene
FADD
Gene Name
Fas associated via death domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003824.3 848 Missense Mutation CAT,CGT H184R NP_003815.1

View Full Product Details