Product Details

SNP ID
rs145644830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:45649981 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATGCCCAGGAACCCGTAGATCT[C/T]CTCGGTCTTCTTCATAGGGTTCCGA
Phenotype
MIM: 603797
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHST1 PubMed Links

Gene Details

Gene
CHST1
Gene Name
carbohydrate sulfotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003654.5 1613 Missense Mutation AAG,GAG K315E NP_003645.1
XM_006718356.3 1613 Missense Mutation AAG,GAG K315E XP_006718419.1
XM_017018459.1 1613 Missense Mutation AAG,GAG K315E XP_016873948.1

View Full Product Details