Product Details
- SNP ID
-
rs145663112
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:76453321 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTATTTTTCTTCATAGCATCTCA[A/G]CAGAACGCCACCGTGCTGAAGTTCG
- Phenotype
-
MIM: 608574
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EMSY
PubMed Links
Gene Details
- Gene
- EMSY
- Gene Name
- EMSY, BRCA2 interacting transcriptional repressor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001300942.1 |
321 |
Missense Mutation |
ACA,GCA |
T60A |
NP_001287871.1 |
NM_001300943.1 |
321 |
Missense Mutation |
ACA,GCA |
T60A |
NP_001287872.1 |
NM_001300944.1 |
321 |
Missense Mutation |
ACA,GCA |
T60A |
NP_001287873.1 |
NM_020193.4 |
321 |
Missense Mutation |
ACA,GCA |
T60A |
NP_064578.2 |
XM_005274112.4 |
321 |
Missense Mutation |
ACA,GCA |
T60A |
XP_005274169.1 |
XM_006718637.3 |
321 |
Missense Mutation |
ACA,GCA |
T60A |
XP_006718700.1 |
XM_006718638.3 |
321 |
Missense Mutation |
ACA,GCA |
T60A |
XP_006718701.1 |
XM_006718639.3 |
321 |
Missense Mutation |
ACA,GCA |
T60A |
XP_006718702.1 |
XM_011545173.2 |
321 |
Missense Mutation |
ACA,GCA |
T60A |
XP_011543475.1 |
XM_011545174.2 |
321 |
Missense Mutation |
ACA,GCA |
T60A |
XP_011543476.1 |
XM_011545175.2 |
321 |
Missense Mutation |
ACA,GCA |
T60A |
XP_011543477.1 |
XM_011545177.2 |
321 |
Missense Mutation |
ACA,GCA |
T60A |
XP_011543479.1 |
XM_011545178.2 |
321 |
Missense Mutation |
ACA,GCA |
T60A |
XP_011543480.1 |
XM_011545179.2 |
321 |
Missense Mutation |
ACA,GCA |
T60A |
XP_011543481.1 |
XM_017018021.1 |
321 |
Missense Mutation |
ACA,GCA |
T60A |
XP_016873510.1 |
XM_017018022.1 |
321 |
UTR 5 |
|
|
XP_016873511.1 |
View Full Product Details