Product Details

SNP ID
rs145693958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:101031678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTGATCATGACATCATTATTTG[A/T]ATACAATGCAGCAAGAATTTTGTTT
Phenotype
MIM: 607311
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PGR PubMed Links

Gene Details

Gene
PGR
Gene Name
progesterone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000926.4 10983 UTR 3 NP_000917.3
NM_001202474.3 10983 UTR 3 NP_001189403.1
NM_001271161.2 10983 UTR 3 NP_001258090.1
NM_001271162.1 10983 UTR 3 NP_001258091.1
XM_006718858.3 10983 Intron XP_006718921.1
XM_011542869.2 10983 Intron XP_011541171.1

View Full Product Details