Product Details

SNP ID
rs145748444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60891253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCGAAGGCCACCCCTGTGGTCAG[C/G]CCGCTATGCTCTGAGGAGAAAGATA
Phenotype
MIM: 608330
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRPF19 PubMed Links

Gene Details

Gene
PRPF19
Gene Name
pre-mRNA processing factor 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014502.4 1635 Silent Mutation GGC,GGG G476G NP_055317.1

View Full Product Details