Product Details

SNP ID
rs145769072
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72229107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAAAGTGCTCCCAATGGGCTGA[A/G]CACCGTCTCGCACGACTACCTGAAA
Phenotype
MIM: 136425 MIM: 600829 MIM: 602753
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOLR2 PubMed Links

Gene Details

Gene
FOLR2
Gene Name
folate receptor beta
There are no transcripts associated with this gene.

Gene
INPPL1
Gene Name
inositol polyphosphate phosphatase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001567.3 907 Missense Mutation AAC,AGC N179S NP_001558.3
XM_005273978.3 907 Missense Mutation AAC,AGC N201S XP_005274035.1
XM_005273979.4 907 Missense Mutation AAC,AGC N201S XP_005274036.1
XM_011544999.2 907 Missense Mutation AAC,AGC N179S XP_011543301.1
XM_011545000.2 907 Missense Mutation AAC,AGC N201S XP_011543302.1
XM_017017673.1 907 Missense Mutation AAC,AGC N191S XP_016873162.1
Gene
PHOX2A
Gene Name
paired like homeobox 2a
There are no transcripts associated with this gene.

View Full Product Details