Product Details

SNP ID
rs145773595
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:121303501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGATTGTAACAAATGTTGGTGC[C/T]TACATCCTTTATTTCTTCTGTGCAA
Phenotype
MIM: 602286
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SC5D PubMed Links

Gene Details

Gene
SC5D
Gene Name
sterol-C5-desaturase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024956.2 347 Silent Mutation GCC,GCT A42A NP_001020127.1
NM_006918.4 347 Silent Mutation GCC,GCT A42A NP_008849.2

View Full Product Details