Product Details

SNP ID
rs145795148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72228479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTGAGCGAGAGCCGGACCCACC[A/G]GATGACCGGGATGCCTCAGGTACTT
Phenotype
MIM: 136425 MIM: 600829
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOLR2 PubMed Links

Gene Details

Gene
FOLR2
Gene Name
folate receptor beta
There are no transcripts associated with this gene.

Gene
INPPL1
Gene Name
inositol polyphosphate phosphatase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001567.3 683 Silent Mutation CCA,CCG P126P NP_001558.3
XM_005273978.3 683 Silent Mutation CCA,CCG P126P XP_005274035.1
XM_005273979.4 683 Silent Mutation CCA,CCG P126P XP_005274036.1
XM_011544999.2 683 Silent Mutation CCA,CCG P126P XP_011543301.1
XM_011545000.2 683 Silent Mutation CCA,CCG P126P XP_011543302.1
XM_017017673.1 683 Silent Mutation CCA,CCG P116P XP_016873162.1

View Full Product Details