Product Details

SNP ID
rs145832034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56093802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATATCTGTCTGGGTGATAAGCAG[G/T]TTGGCGTTCTGTGATTCCAGCATCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR8I2 PubMed Links

Gene Details

Gene
OR8I2
Gene Name
olfactory receptor family 8 subfamily I member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003750.1 Intron NP_001003750.1

View Full Product Details