Product Details

SNP ID
rs145833926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:134332137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTGGTCGTCTTGGGCTTCCTG[A/G]TGCTCCGCAGGTGAGAGAGAGCTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLB1L2 PubMed Links

Gene Details

Gene
GLB1L2
Gene Name
galactosidase beta 1 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138342.3 264 Missense Mutation ATG,GTG M26V NP_612351.2
XM_006718937.1 264 Missense Mutation ATG,GTG M26V XP_006719000.1

View Full Product Details