Product Details

SNP ID
rs145860973
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5422510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGAGGCCTGTTTTGCTCAGTTT[G/T]TCTTCCTTCATGGATTCTCCTTTAT
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
OR51B5 PubMed Links
Additional Information
For this assay, SNP(s) [rs2736587] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
OR51B5
Gene Name
olfactory receptor family 51 subfamily B member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005567.2 400 Intron NP_001005567.2
Gene
OR51Q1
Gene Name
olfactory receptor family 51 subfamily Q member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004757.2 400 Missense Mutation GTC,TTC V104F NP_001004757.1

View Full Product Details