Product Details

SNP ID
rs145991257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:86306344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGTGGTTTTTATGCTGGGAACA[A/G]TCCCATTTCCTGAGGGAATGGGAGG
Phenotype
MIM: 614908
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIKESHI PubMed Links

Gene Details

Gene
HIKESHI
Gene Name
Hikeshi, heat shock protein nuclear import factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322404.1 145 Missense Mutation ATC,GTC I44V NP_001309333.1
NM_001322407.1 145 Missense Mutation ATC,GTC I5V NP_001309336.1
NM_001322409.1 145 Missense Mutation ATC,GTC I5V NP_001309338.1
NM_016401.3 145 Missense Mutation ATC,GTC I44V NP_057485.2
XM_017017914.1 145 Missense Mutation ATC,GTC I44V XP_016873403.1
XM_017017915.1 145 Missense Mutation ATC,GTC I5V XP_016873404.1

View Full Product Details