Product Details

SNP ID
rs146030294
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66332453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACCCTTGCTCCTCCCCTCTGCT[C/T]CTTGCCTCTGACTGCCCACTGCCCT
Phenotype
MIM: 606259 MIM: 605965
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
BRMS1 PubMed Links
Additional Information
For this assay, SNP(s) [rs1784029] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
BRMS1
Gene Name
breast cancer metastasis suppressor 1
There are no transcripts associated with this gene.

Gene
RIN1
Gene Name
Ras and Rab interactor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004292.2 2651 Silent Mutation AGA,AGG R725R NP_004283.2
XM_011545399.2 2651 Silent Mutation AGA,AGG R697R XP_011543701.1
XM_011545400.2 2651 Silent Mutation AGA,AGG R663R XP_011543702.1
XM_011545401.2 2651 Silent Mutation AGA,AGG R644R XP_011543703.1
XM_017018587.1 2651 Silent Mutation AGA,AGG R664R XP_016874076.1

View Full Product Details