Product Details

SNP ID
rs146157876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:113904948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCCCAGGAATGTTGTCAAGTGT[A/T]ATGGCTCCCCTGTGGGCCTGCATCC
Phenotype
MIM: 604654
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HTR3B PubMed Links

Gene Details

Gene
HTR3B
Gene Name
5-hydroxytryptamine receptor 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006028.4 153 Silent Mutation GTA,GTT V5V NP_006019.1
XM_011543063.1 153 Intron XP_011541365.1
XM_017018552.1 153 Intron XP_016874041.1

View Full Product Details