Product Details

SNP ID
rs146213665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1840872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGATGCGGCTGAAGAGGAGAAGTA[C/T]GACATGGAGGTGAGGGTGCAGAAGA
Phenotype
MIM: 607719 MIM: 191043
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SYT8 PubMed Links

Gene Details

Gene
SYT8
Gene Name
synaptotagmin 8
There are no transcripts associated with this gene.

Gene
TNNI2
Gene Name
troponin I2, fast skeletal type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145829.1 262 Silent Mutation TAC,TAT Y80Y NP_001139301.1
NM_001145841.1 262 Silent Mutation TAC,TAT Y80Y NP_001139313.1
NM_003282.3 262 Silent Mutation TAC,TAT Y80Y NP_003273.1

View Full Product Details