Product Details

SNP ID
rs146234178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4999114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATATTTGGTAGCATTTATGGGTAA[C/T]GTTACCATCCTGTCTGTCATTTGGA
Phenotype
MIM: 605470
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MMP26 PubMed Links

Gene Details

Gene
MMP26
Gene Name
matrix metallopeptidase 26
There are no transcripts associated with this gene.

Gene
OR51L1
Gene Name
olfactory receptor family 51 subfamily L member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004755.1 132 Silent Mutation AAC,AAT N44N NP_001004755.1

View Full Product Details