Product Details

SNP ID
rs146268368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:33090610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGTATCTATCTACTAGTAAAGC[C/T]GTTTCTTTCCCTTCATACTCTTCTT
Phenotype
MIM: 600367
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSTF3 PubMed Links

Gene Details

Gene
CSTF3
Gene Name
cleavage stimulation factor subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033505.1 1729 Intron NP_001028677.1
NM_001033506.1 1729 Intron NP_001028678.1
NM_001326.2 1729 Silent Mutation ACA,ACG T521T NP_001317.1

View Full Product Details