Product Details

SNP ID
rs146355768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64730757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGACACAGACCTCAGTTCAAATC[A/G]CAGCTCTACTTCCCTGCCTCTTGAC
Phenotype
MIM: 600566 MIM: 605577
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NRXN2 PubMed Links

Gene Details

Gene
NRXN2
Gene Name
neurexin 2
There are no transcripts associated with this gene.

Gene
RASGRP2
Gene Name
RAS guanyl releasing protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098670.1 Intron NP_001092140.1
NM_001098671.1 Intron NP_001092141.1
NM_001318398.1 Intron NP_001305327.1
NM_153819.1 Intron NP_722541.1
XM_005273707.3 Intron XP_005273764.3
XM_011544718.1 Intron XP_011543020.1
XM_011544720.1 Intron XP_011543022.1
XM_011544721.1 Intron XP_011543023.1
XM_011544722.1 Intron XP_011543024.1
XM_011544723.2 Intron XP_011543025.1
XM_011544725.2 Intron XP_011543027.1
XM_017017082.1 Intron XP_016872571.1
XM_017017083.1 Intron XP_016872572.1
XM_017017084.1 Intron XP_016872573.1
XM_017017085.1 Intron XP_016872574.1
XM_017017086.1 Intron XP_016872575.1

View Full Product Details