Product Details

SNP ID
rs146538294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6319353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCGGGGTGGGCGGTGGCGCTGCA[A/G]GGCCTTTCCGGCCCGAAAGGGCCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRKCDBP PubMed Links

Gene Details

Gene
PRKCDBP
Gene Name
protein kinase C delta binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145040.2 630 Missense Mutation CCT,CTT P199L NP_659477.2

View Full Product Details