Product Details

SNP ID
rs146593182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68003029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGCAGCCGCGCCCACCTGATGA[G/T]CACAGGTGTGTAGAGCAGGGCGGCG
Phenotype
MIM: 600466 MIM: 608204
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ALDH3B1 PubMed Links

Gene Details

Gene
ALDH3B1
Gene Name
aldehyde dehydrogenase 3 family member B1
There are no transcripts associated with this gene.

Gene
UNC93B1
Gene Name
unc-93 homolog B1 (C. elegans)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030930.3 76 Missense Mutation ATC,CTC I129L NP_112192.2
XM_011545290.1 76 UTR 5 XP_011543592.1
XM_011545291.2 76 UTR 5 XP_011543593.1

View Full Product Details