Product Details
- SNP ID
-
rs146648668
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:66314867 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCAGCGATAGCAGTCAGTGATGC[A/G]CTTGTTGGGTGCATGGGGGCCACAG
- Phenotype
-
MIM: 606064
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD248
PubMed Links
Gene Details
- Gene
- CD248
- Gene Name
- CD248 molecule
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020404.2 |
2178 |
Missense Mutation |
CGC,TGC |
R721C |
NP_065137.1 |
View Full Product Details