Product Details

SNP ID
rs146680546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59806653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGCCAGCCTTCACAGGTGTCAC[A/G]TAGTGGTCAATAGCACCTTTGCCTC
Phenotype
MIM: 611829 MIM: 600876
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL16 PubMed Links

Gene Details

Gene
MRPL16
Gene Name
mitochondrial ribosomal protein L16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017840.3 664 Silent Mutation TAC,TAT Y150Y NP_060310.1
Gene
STX3
Gene Name
syntaxin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178040.1 664 Intron NP_001171511.1
NM_004177.4 664 Intron NP_004168.1
XM_005274195.3 664 Intron XP_005274252.1
XM_005274198.2 664 Intron XP_005274255.1
XM_005274200.3 664 Intron XP_005274257.1
XM_011545221.2 664 Intron XP_011543523.1
XM_017018188.1 664 Intron XP_016873677.1
XM_017018189.1 664 Intron XP_016873678.1
XM_017018190.1 664 Intron XP_016873679.1
XM_017018191.1 664 Intron XP_016873680.1
XM_017018192.1 664 Intron XP_016873681.1
XM_017018193.1 664 Intron XP_016873682.1
XM_017018194.1 664 Intron XP_016873683.1
XM_017018195.1 664 Intron XP_016873684.1
XM_017018196.1 664 Intron XP_016873685.1

View Full Product Details