Product Details

SNP ID
rs146744565
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59835884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCACTTTTCACACTAACATTGA[C/T]GGTCTCGTTGAAGAGCAGCTCAACC
Phenotype
MIM: 609342
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GIF PubMed Links

Gene Details

Gene
GIF
Gene Name
gastric intrinsic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005142.2 1031 Missense Mutation ATC,GTC I333V NP_005133.2
XM_011544939.2 1031 Missense Mutation ATC,GTC I319V XP_011543241.1

View Full Product Details