Product Details

SNP ID
rs146760101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68065030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATCTCAGGAGGAAATTCCCATGA[A/G]AACTGATTCTCCACACAATCACCAG
Phenotype
MIM: 118491
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHKA PubMed Links

Gene Details

Gene
CHKA
Gene Name
choline kinase alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277.2 Intron NP_001268.2
NM_212469.1 Intron NP_997634.1
XM_017017147.1 Intron XP_016872636.1
XM_017017148.1 Intron XP_016872637.1

View Full Product Details