Product Details

SNP ID
rs146840475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72687717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTTCTCATGTTTCTCTGTCTC[A/G]TGCACCACTGTGAAGCCCCAGCTAC
Phenotype
MIM: 606646
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARAP1 PubMed Links

Gene Details

Gene
ARAP1
Gene Name
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040118.2 4092 Silent Mutation CAC,CAT H1364H NP_001035207.1
NM_001135190.1 4092 Silent Mutation CAC,CAT H1047H NP_001128662.1
NM_015242.4 4092 Silent Mutation CAC,CAT H1119H NP_056057.2

View Full Product Details