Product Details

SNP ID
rs146879673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65867016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTGACCATCTCCAGGTCCAGCA[C/G]GTACTCCCGGGGGCCCGTCACCGGC
Phenotype
MIM: 601442 MIM: 604633 MIM: 606591
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CFL1 PubMed Links

Gene Details

Gene
CFL1
Gene Name
cofilin 1
There are no transcripts associated with this gene.

Gene
EFEMP2
Gene Name
EGF containing fibulin like extracellular matrix protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016938.4 1493 Missense Mutation CTG,GTG L412V NP_058634.4
Gene
MUS81
Gene Name
MUS81 structure-specific endonuclease subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025128.4 1493 Intron NP_079404.3
XM_005274307.2 1493 Intron XP_005274364.1
XM_011545269.1 1493 Intron XP_011543571.1
XM_011545270.1 1493 Intron XP_011543572.1

View Full Product Details