Product Details

SNP ID
rs147162223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102790725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCATCTTTCATGAAAACTGCATC[A/G]ACTTTGTGGCCAATTCCAGGAAAGT
Phenotype
MIM: 120353
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MMP1 PubMed Links

Gene Details

Gene
MMP1
Gene Name
matrix metallopeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145938.1 1421 Silent Mutation GTC,GTT V360V NP_001139410.1
NM_002421.3 1421 Silent Mutation GTC,GTT V426V NP_002412.1
Gene
WTAPP1
Gene Name
Wilms tumor 1 associated protein pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details