Product Details

SNP ID
rs147240748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118660623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAACAGGCACCAGTTTGCTTGTTC[A/G]GATGCCGCTAAGCGAGTTGGGGTTT
Phenotype
MIM: 612834 MIM: 275360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHLDB1 PubMed Links

Gene Details

Gene
PHLDB1
Gene Name
pleckstrin homology like domain family B member 1
There are no transcripts associated with this gene.

Gene
TREH
Gene Name
trehalase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301065.1 1063 Nonsense Mutation CGA,TGA R309* NP_001287994.1
NM_007180.2 1063 Nonsense Mutation CGA,TGA R340* NP_009111.2

View Full Product Details