Product Details
- SNP ID
-
rs147545257
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:59806526 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTTTCGCATCTTCTCTAGAGTCC[C/T]GCGGCTCACAGCCTTTGCTGCGAAG
- Phenotype
-
MIM: 611829
MIM: 600876
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MRPL16
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs491671] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MRPL16
- Gene Name
- mitochondrial ribosomal protein L16
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017840.3 |
791 |
Missense Mutation |
AGG,GGG |
R193G |
NP_060310.1 |
- Gene
- STX3
- Gene Name
- syntaxin 3
View Full Product Details