Product Details

SNP ID
rs147645347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:639516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTCATGCTGTGCACCGCCTCCAT[C/T]TTCAACCTGTGCGCCATCAGCGTGG
Phenotype
MIM: 602635 MIM: 126452
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DEAF1 PubMed Links

Gene Details

Gene
DEAF1
Gene Name
DEAF1, transcription factor
There are no transcripts associated with this gene.

Gene
DRD4
Gene Name
dopamine receptor D4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000797.3 369 Silent Mutation ATC,ATT I123I NP_000788.2

View Full Product Details