Product Details

SNP ID
rs147668114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6169404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCAAGTCTTGATGTCAAAGAAC[A/C]GGTGGGCTACACCCTCACGTATCTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR52B2 PubMed Links

Gene Details

Gene
OR52B2
Gene Name
olfactory receptor family 52 subfamily B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004052.1 923 Missense Mutation NP_001004052.1

View Full Product Details