Product Details

SNP ID
rs147883248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62615779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCTTCATCTTGGGCTTCTCTGT[C/T]CGAGTATGCCACACCAGTACCTGTG
Phenotype
MIM: 606374 MIM: 104160 MIM: 180721
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B3GAT3 PubMed Links

Gene Details

Gene
B3GAT3
Gene Name
beta-1,3-glucuronyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288721.1 1595 Silent Mutation CGA,CGG R303R NP_001275650.1
NM_001288722.1 1595 UTR 3 NP_001275651.1
NM_001288723.1 1595 UTR 3 NP_001275652.1
NM_012200.3 1595 Silent Mutation CGA,CGG R310R NP_036332.2
Gene
EML3
Gene Name
echinoderm microtubule associated protein like 3
There are no transcripts associated with this gene.

Gene
GANAB
Gene Name
glucosidase II alpha subunit
There are no transcripts associated with this gene.

Gene
ROM1
Gene Name
retinal outer segment membrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000327.3 1595 Intron NP_000318.1

View Full Product Details