Product Details

SNP ID
rs147914844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64935358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTCCGGTCCCAGAGGTACTCAC[A/G]GTGCAAGTGGCAGCCTGGGATGACT
Phenotype
MIM: 609651 MIM: 601644
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPHA2 PubMed Links

Gene Details

Gene
GPHA2
Gene Name
glycoprotein hormone alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130769.3 903 Missense Mutation CCC,TCC P35S NP_570125.1
XM_011544774.2 903 Missense Mutation CCC,TCC P68S XP_011543076.1
XM_011544775.2 903 Missense Mutation CCC,TCC P35S XP_011543077.1
XM_011544776.2 903 Missense Mutation CCC,TCC P35S XP_011543078.1
Gene
MAJIN
Gene Name
membrane anchored junction protein
There are no transcripts associated with this gene.

Gene
PPP2R5B
Gene Name
protein phosphatase 2 regulatory subunit B'beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006244.3 903 Intron NP_006235.1
XM_011545132.2 903 Intron XP_011543434.1
XM_011545133.1 903 Intron XP_011543435.1
XM_011545134.2 903 Intron XP_011543436.1

View Full Product Details