Product Details

SNP ID
rs147988588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:821757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTGAAGGTCCTGCCTGCTGATA[C/G]CCATGAGCATGCCAGTGGGCGCCTG
Phenotype
MIM: 614177 MIM: 609059 MIM: 180530
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CRACR2B PubMed Links

Gene Details

Gene
CRACR2B
Gene Name
calcium release activated channel regulator 2B
There are no transcripts associated with this gene.

Gene
PNPLA2
Gene Name
patatin like phospholipase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020376.3 466 Missense Mutation ACC,AGC T106S NP_065109.1
XM_006718265.3 466 Missense Mutation ACC,AGC T106S XP_006718328.1
XM_006718266.3 466 Missense Mutation ACC,AGC T106S XP_006718329.1
XM_017018028.1 466 Missense Mutation ACC,AGC T106S XP_016873517.1
Gene
RPLP2
Gene Name
ribosomal protein lateral stalk subunit P2
There are no transcripts associated with this gene.

Gene
SNORA52
Gene Name
small nucleolar RNA, H/ACA box 52
There are no transcripts associated with this gene.

View Full Product Details