Product Details

SNP ID
rs147995731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4571458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTCAAGTAGTTCACATCCACAT[G/T]AAGTAGAACACGATCCATACAGTTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C11orf40 PubMed Links

Gene Details

Gene
C11orf40
Gene Name
chromosome 11 open reading frame 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144663.1 619 Missense Mutation AAT,CAT N207H NP_653264.1

View Full Product Details