Product Details

SNP ID
rs78255170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4955705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTTCAACATATGATGTGTTGAT[A/G]ATGGACATGATTCATTCATAGGGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR51A2 PubMed Links

Gene Details

Gene
OR51A2
Gene Name
olfactory receptor family 51 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004748.1 9 Silent Mutation ATC,ATT I3I NP_001004748.1
Gene
OR51A4
Gene Name
olfactory receptor family 51 subfamily A member 4
There are no transcripts associated with this gene.

View Full Product Details