Product Details

SNP ID
rs148078235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47441692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCCCAGGCGGTTTCCGATCTC[A/G]GTCATGATGCTCATGGCGGAGTCGT
Phenotype
MIM: 601592
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAPSN PubMed Links

Gene Details

Gene
RAPSN
Gene Name
receptor associated protein of the synapse
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005055.4 1045 Silent Mutation ACC,ACT T277T NP_005046.2
NM_032645.4 1045 Intron NP_116034.2
XM_005253042.3 1045 Silent Mutation ACC,ACT T277T XP_005253099.1
XM_005253043.3 1045 Intron XP_005253100.1
XM_011520252.1 1045 Silent Mutation ACC,ACT T277T XP_011518554.1

View Full Product Details